Learn More
Medchemexpress LLC CPD-10 | 3055471-57-0 | 96.7% | 888.07 | 1 MG
SDP
Catalog No. 5000390519
Encompass_Preferred
CPD-10 is a potent CCND1 and CDK4 PROTAC degrader. It shows anti-proliferation and induces apoptosis. CPD-10 decreases the protein expression of cyclin D1, cyclin D3, CDK4, and P-Rb(5807/811) in a dose-dependent manner.
  • Potent CCND1 and CDK4 PROTAC degrader
  • Shows anti-proliferation
  • Induces apoptosis
  • Decreases protein expression of cyclin D1, cyclin D3, CDK4, P-Rb(5807/811) in a dose-dependent manner
Catalog No. 50-003-90519 Supplier Medchemexpress LLC Supplier No. HY1686601MG
CPD-10 is a potent CCND1 and CDK4 PROTAC degrader. It shows anti-proliferation and induces apoptosis. CPD-10 decreases the protein expression of cyclin D1, cyclin D3, CDK4, and P-Rb(5807/811) in a dose-dependent manner.
  • Potent CCND1 and CDK4 PROTAC degrader
  • Shows anti-proliferation
  • Induces apoptosis
  • Decreases protein expression of cyclin D1, cyclin D3, CDK4, P-Rb(5807/811) in a dose-dependent manner