Learn More
Medchemexpress LLC Cpd-10 | 3055471-57-0 | 96.7% | 888.07 | 50 MG
SDP
Catalog No. 5000391059
Encompass_Preferred
CPD-10 is a potent CCND1 and CDK4 PROTAC degrader that exhibits anti-proliferation effects and induces apoptosis. It decreases the protein expression of cyclin D1, cyclin D3, CDK4, and P-Rb(5807/811) in a dose-dependent manner. This compound is composed of a CDK ligand, an E3 Ligase Ligand-Linker Conjugates ligand, and a linker.
  • Potent CCND1 and CDK4 PROTAC degrader
  • Shows anti-proliferation
  • Induces apoptosis
  • Decreases protein expression of cyclin D1, cyclin D3, CDK4, P-Rb(5807/811) in a dose-dependent manner
Catalog No. 50-003-91059 Supplier Medchemexpress LLC Supplier No. HY16866050MG
CPD-10 is a potent CCND1 and CDK4 PROTAC degrader that exhibits anti-proliferation effects and induces apoptosis. It decreases the protein expression of cyclin D1, cyclin D3, CDK4, and P-Rb(5807/811) in a dose-dependent manner. This compound is composed of a CDK ligand, an E3 Ligase Ligand-Linker Conjugates ligand, and a linker.
  • Potent CCND1 and CDK4 PROTAC degrader
  • Shows anti-proliferation
  • Induces apoptosis
  • Decreases protein expression of cyclin D1, cyclin D3, CDK4, P-Rb(5807/811) in a dose-dependent manner