Learn More
Medchemexpress LLC CPD-1224 | 2891620-68-9 | C43H47ClN8O7S | 100 MG
SDP
Catalog No. 5000373243
Encompass_Preferred
CPD-1224 is an orally effective ALK inhibitor derived from an ALK inhibitor that connects to the cereblon ligand. It targets the EML4-ALK oncogenic fusion protein, leading to the degradation of both ALK and its mutant forms L1196M/G1202R. This compound has the potential to slow down tumor growth and is for research use only.
  • Enhances kinase binding selectivity and exhibits good degradation selectivity
  • Shows antiproliferative activity against mouse Ba/F3 cells
  • Demonstrates oral bioavailability of 28% in mice (10 mg/kg, oral)
  • Inhibits tumor growth in mice by reducing total ALK and phosphorylated ALK levels
Catalog No. 50-003-73243 Supplier Medchemexpress LLC Supplier No. HY153386100MG
CPD-1224 is an orally effective ALK inhibitor derived from an ALK inhibitor that connects to the cereblon ligand. It targets the EML4-ALK oncogenic fusion protein, leading to the degradation of both ALK and its mutant forms L1196M/G1202R. This compound has the potential to slow down tumor growth and is for research use only.
  • Enhances kinase binding selectivity and exhibits good degradation selectivity
  • Shows antiproliferative activity against mouse Ba/F3 cells
  • Demonstrates oral bioavailability of 28% in mice (10 mg/kg, oral)
  • Inhibits tumor growth in mice by reducing total ALK and phosphorylated ALK levels