Learn More
Medchemexpress LLC HY-18685 10mg Medchemexpress, CPDA CAS:1415834-63-7 Purity:>98%
SDP
Catalog No. 502082070
Encompass_Preferred
Medchemexpress, HY-18685 10mg CPDA CAS:1415834-63-7 CPDA is a novel potent SH2 domain-containing inositol phosphatase 2 (SHIP2) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2070 Supplier Medchemexpress LLC Supplier No. HY1868510MG
Medchemexpress, HY-18685 10mg CPDA CAS:1415834-63-7 CPDA is a novel potent SH2 domain-containing inositol phosphatase 2 (SHIP2) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.