Learn More
Aviva Systems Biology Corp CPE Antibody - N-terminal region (ARP58445_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58445_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CPE. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62310