Learn More
VGuard™ CPE Shoe Covers
Catalog No. 17005971
Click to view available options
Color:
Blue
White
Size:
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
17-005-971 White Large
17-005-972 White X-Large
17-005-974 Blue X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-005-971 Supplier VGuard™ Supplier No. B95L13
Add to Cart
Add to Cart
This item is not returnable. View return policy
Durable, embossed CPE shoe covers provides traction on slippery surfaces, while protecting shoes and surfaces from fluids, grease, oil, dirt, and grime.
  • Disposable
  • Elastic top fits snugly for easy donning
  • Available in white or blue. Blue color style aids in contamination control
Specifications
Size Large
Material Cast Polyethylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Non-skid
Model B95L13
Certifications/Compliance 21 CFR, Parts 170-199