Learn More
Aviva Systems Biology Corp CPEB2 Antibody - middle region (ARP41185_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41185_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPEB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CPEB2. Alternate names: CPEB-2, CPE-BP2, hCPEB-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53285