Learn More
ABclonal Technology CPEB3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A154021000UL
Encompass_Preferred
Enables mRNA 3-UTR binding activity and translation factor activity, RNA binding. Involved in cellular response to amino acid stimulus, negative regulation of transcription by RNA polymerase II, and positive regulation of mRNA catabolic process. Located in several cellular components, including cytosol, midbody, and nucleoplasm. Part of CCR4-NOT complex.
Catalog No. 50-001-07626