Learn More
Aviva Systems Biology Corp CPEB4 Antibody - N-terminal region (ARP41024_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41024_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPEB4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CPEB4. Alternate names: KIAA1673ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-53262