Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ CPG (Controlled Pore Glass) Synthesis Column, 500 Angstrom, 10.0 μmol, dC(Bz)

Catalog No. 400391
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Column
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-039-1 1 Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-039-1 Supplier Applied Biosystems™ Supplier No. 400391
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 10.0 μmol dC(Bz) Controlled Pore Glass (CPG) columns are suitable for the synthesis of oligonucleotides <50 bases in length.

The 10.0 μmol dC(Bz) Controlled Pore Glass (CPG) columns are suitable for the synthesis of oligonucleotides <50 bases in length. These columns deliver high-quality oligonucleotides.

  • Proprietary design and manufacturing methods ensure consistency and high quality
  • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
  • The 500 Å pore size allows synthesis of oligonucleotides of up to 50 bases.

These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.

TRUSTED_SUSTAINABILITY

Specifications

Column Type Oligonucleotide Synthesis
Content And Storage Columns may be stored in a dry environment at room temperature.
Format Column
Sample Type Oligos
For Use With (Equipment) 3900 DNA Synthesizer
For Use With (Application) Oligo Synthesis
High-throughput Compatibility Not High-throughput Compatible (Manual)
Matrix Controlled Pore Glass
Pore Size 500 Å
Prep Scale 10 μmol
Product Type Oligonucleotide Synthesis Columns
Quantity 1 Column
Shipping Condition Room Temperature
Target Nucleic Acids
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.