Learn More
New England Biolabs, Inc. CpG Methyltransferase (M.SssI) – 100 units
SDP
Catalog No. 50811649
Encompass
The CpG Methyltransferase, M.SssI, methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5'...CG...3'
Catalog No. 50-811-649 Supplier New England Biolabs, Inc. Supplier No. M0226S
The CpG Methyltransferase, M.SssI, methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5'...CG...3'