Learn More
New England Biolabs, Inc. CpG Methyltransferase (M.SssI) – 500 units
SDP
Catalog No. 50811648
Encompass
The CpG Methyltransferase, M.SssI, methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5'...CG...3'
Catalog No. 50-811-648 Supplier New England Biolabs, Inc. Supplier No. M0226M
The CpG Methyltransferase, M.SssI, methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5'...CG...3'