Learn More
Zymo Research Corporation CpG Methylase (M.SssI), 200 Units
SDP
Catalog No. 502037191
Encompass
The recombinant CpG Methylase is isolated from an E. coli strain that expresses the methyltransferase gene from Spiroplasma sp. strain MQ1. The reaction conditions are optimized to maximize the processivity of the enzyme to ensure rapid, complete, and reproducible methylation of DNA for accurate DNA methylation analysis.
Catalog No. 50-203-7191 Supplier Zymo Research Corporation Supplier No. E2010
The recombinant CpG Methylase is isolated from an E. coli strain that expresses the methyltransferase gene from Spiroplasma sp. strain MQ1. The reaction conditions are optimized to maximize the processivity of the enzyme to ensure rapid, complete, and reproducible methylation of DNA for accurate DNA methylation analysis.