Learn More
Medchemexpress LLC CpG ODN 10101 | 1234402-77-7 | DNA, d(P-thio)(T-C-G-T-C-G-T-T-T-T-C-G-C-G-C-G-C-G-C-C-G) | 1 MG
SDP
Catalog No. 5000375351
Encompass_Preferred
CpG ODN 10101 is a selective agonist targeting TLR9, a synthetic oligodeoxynucleotide modified with phosphate thioester. It activates B cells and plasmacytoid dendritic cells (pDCs), inducing the production of various cytokines and chemokines. This regulates innate immunity and promotes Th1 adaptive immune responses, making it suitable as an immunomodulator and vaccine adjuvant.
  • Selective agonist targeting TLR9
  • Synthetic oligodeoxynucleotide modified with phosphate thioester
  • Activates B cells and plasmacytoid dendritic cells (pDCs)
  • Induces cytokine and chemokine production
  • Regulates innate immunity and promotes Th1 adaptive immune responses
  • Possesses antiviral properties
  • Enhances vaccine immunogenicity
  • Suitable as an immunomodulator and vaccine adjuvant
Catalog No. 50-003-75351 Supplier Medchemexpress LLC Supplier No. HY1502171MG
CpG ODN 10101 is a selective agonist targeting TLR9, a synthetic oligodeoxynucleotide modified with phosphate thioester. It activates B cells and plasmacytoid dendritic cells (pDCs), inducing the production of various cytokines and chemokines. This regulates innate immunity and promotes Th1 adaptive immune responses, making it suitable as an immunomodulator and vaccine adjuvant.
  • Selective agonist targeting TLR9
  • Synthetic oligodeoxynucleotide modified with phosphate thioester
  • Activates B cells and plasmacytoid dendritic cells (pDCs)
  • Induces cytokine and chemokine production
  • Regulates innate immunity and promotes Th1 adaptive immune responses
  • Possesses antiviral properties
  • Enhances vaccine immunogenicity
  • Suitable as an immunomodulator and vaccine adjuvant