Learn More
TARGETMOL CHEMICALS INC CpG ODN 10101 1MG
SDP
Catalog No. 502876226
Encompass_Preferred
Also available in 5 mg and bulk. Please contact Fisher for quotes. CpG ODN 10101 (CPG 10101) is a novel potent and selective Toll-like receptor 9 (TLR9)-containing oligodeoxynucleotide agonist with antiviral activity for the treatment of chronic infections. Purity 94.06%
Catalog No. 50-287-6226 Supplier TARGETMOL CHEMICALS INC Supplier No. T748741MG
Also available in 5 mg and bulk. Please contact Fisher for quotes. CpG ODN 10101 (CPG 10101) is a novel potent and selective Toll-like receptor 9 (TLR9)-containing oligodeoxynucleotide agonist with antiviral activity for the treatment of chronic infections. Purity 94.06%