Learn More
Medchemexpress LLC CpG ODN 10101 (sodium) | 1234402-77-7 | 99.2% | 7,185.00 Da | 10 MG
SDP
Catalog No. 5000286204
Encompass_Preferred
CpG ODN 10101 (sodium) is a synthetic oligodeoxynucleotide that functions as a toll-like receptor 9 (TLR9) agonist. Supplied as the sodium salt for research use, it is employed to stimulate innate immune responses and study immunomodulatory effects, including cytokine production and B-cell activation.
  • TLR9 agonist that stimulates dendritic cells and B cells.
  • Induces interferon and proinflammatory cytokine production.
  • Provided as a sodium salt to aid stability and handling.
  • High purity suitable for research applications.
  • Available in milligram-scale quantities for laboratory studies.
Catalog No. 50-002-86204 Supplier Medchemexpress LLC Supplier No. HY150217A10MG
CpG ODN 10101 (sodium) is a synthetic oligodeoxynucleotide that functions as a toll-like receptor 9 (TLR9) agonist. Supplied as the sodium salt for research use, it is employed to stimulate innate immune responses and study immunomodulatory effects, including cytokine production and B-cell activation.
  • TLR9 agonist that stimulates dendritic cells and B cells.
  • Induces interferon and proinflammatory cytokine production.
  • Provided as a sodium salt to aid stability and handling.
  • High purity suitable for research applications.
  • Available in milligram-scale quantities for laboratory studies.