Learn More
Medchemexpress LLC CPI-1189 | 183619-38-7 | 99.8% | 234.29 g/mol | C13H18N2O2 | 100 MG
SDP
Catalog No. 5000214214
Encompass_Preferred
CPI-1189 is an orally active TNF-α release inhibitor that suppresses p38 MAP kinase phosphorylation and has demonstrated anti-apoptotic activity. It is used in preclinical research on inflammatory signaling, HIV, and neurological disease models.
  • Orally active TNF-α release inhibitor suitable for in vitro and in vivo studies.
  • Inhibits p38 phosphorylation, useful for signaling pathway research.
  • Demonstrated anti-apoptotic activity in relevant models.
  • High purity (99.82%) solid, white to off-white powder.
  • Stable when stored at -20°C as powder and under recommended solvent conditions.
  • Available in small research pack sizes to support dose-ranging experiments.
Catalog No. 50-002-14214 Supplier Medchemexpress LLC Supplier No. HY100376100MG
CPI-1189 is an orally active TNF-α release inhibitor that suppresses p38 MAP kinase phosphorylation and has demonstrated anti-apoptotic activity. It is used in preclinical research on inflammatory signaling, HIV, and neurological disease models.
  • Orally active TNF-α release inhibitor suitable for in vitro and in vivo studies.
  • Inhibits p38 phosphorylation, useful for signaling pathway research.
  • Demonstrated anti-apoptotic activity in relevant models.
  • High purity (99.82%) solid, white to off-white powder.
  • Stable when stored at -20°C as powder and under recommended solvent conditions.
  • Available in small research pack sizes to support dose-ranging experiments.