Learn More
Medchemexpress LLC Cpi-1612 | 2374971-81-8 | 99.7% | 450.53 g·mol-1 | C27H26N6O | 5 MG
SDP
Catalog No. 5000222495
Encompass_Preferred
CPI-1612 is a potent, orally bioavailable inhibitor of the EP300/CBP histone acetyltransferase (HAT) family used as a research tool in epigenetics and oncology studies. It shows low-nanomolar biochemical potency and cellular activity, and is supplied as a powder for laboratory use.
  • Potent EP300/CBP HAT inhibition (IC50 8.1 nM for EP300 HAT).
  • Orally bioavailable with demonstrated in vivo activity.
  • Cellular activity in cancer cell lines, supporting translational studies.
  • Supplied as a powder with recommended low-temperature storage.
  • Molecular formula C27H26N6O; molecular weight 450.53 g·mol-1.
Catalog No. 50-002-22495 Supplier Medchemexpress LLC Supplier No. HY1362855MG
CPI-1612 is a potent, orally bioavailable inhibitor of the EP300/CBP histone acetyltransferase (HAT) family used as a research tool in epigenetics and oncology studies. It shows low-nanomolar biochemical potency and cellular activity, and is supplied as a powder for laboratory use.
  • Potent EP300/CBP HAT inhibition (IC50 8.1 nM for EP300 HAT).
  • Orally bioavailable with demonstrated in vivo activity.
  • Cellular activity in cancer cell lines, supporting translational studies.
  • Supplied as a powder with recommended low-temperature storage.
  • Molecular formula C27H26N6O; molecular weight 450.53 g·mol-1.