Learn More
Medchemexpress LLC CPI-1612 | 2374971-81-8 | 99.8% | 450.53 | 100 MG
SDP
Catalog No. 5000348765
Encompass_Preferred
CPI-1612 is a highly potent, orally active EP300/CBP histone acetyltransferase (HAT) inhibitor. It exhibits an IC50 of 8.1 nM for EP300 HAT and demonstrates anticancer activity. This product is intended for research use only.
  • Inhibits full length EP300 with an IC50 of <0.5 nM.
  • Inhibits full length CBP with an IC50 of 2.9 nM.
  • Reduces H3K18Ac MSD and JEKO-1 cell proliferation.
  • Shows tumor growth inhibition in animal models.
  • Exhibits high brain penetrance in CD-1 mice.
Catalog No. 50-003-48765 Supplier Medchemexpress LLC Supplier No. HY136285100MG
CPI-1612 is a highly potent, orally active EP300/CBP histone acetyltransferase (HAT) inhibitor. It exhibits an IC50 of 8.1 nM for EP300 HAT and demonstrates anticancer activity. This product is intended for research use only.
  • Inhibits full length EP300 with an IC50 of <0.5 nM.
  • Inhibits full length CBP with an IC50 of 2.9 nM.
  • Reduces H3K18Ac MSD and JEKO-1 cell proliferation.
  • Shows tumor growth inhibition in animal models.
  • Exhibits high brain penetrance in CD-1 mice.