Learn More
Apexbio Technology LLC CPI-169 1450655-76-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063785
Encompass_Preferred
CPI-169 is a selective small-molecule inhibitor targeting enhancer of zeste homolog 2 (EZH2) a catalytic component of polycomb repressive complex 2 (PRC2) By inhibiting EZH2 enzymatic activity CPI-169 suppresses methylation at histone H3 lysine 27 (H3K27me3) thereby altering chromatin structure and gene expression profiles Pharmacological experiments demonstrate inhibition of EZH2 with subnanomolar IC50 values and cellular studies indicate reductions of H3K27 trimethylation at nanomolar concentrations CPI-169 has been utilized experimentally to investigate EZH2-dependent epigenetic processes cell proliferation apoptosis pathways and therapeutic potential particularly in lymphoma and cancer biology studies employing in vitro and in vivo models
Catalog No. 50-000-63785 Supplier Apexbio Technology LLC Supplier No. B46785.1
CPI-169 is a selective small-molecule inhibitor targeting enhancer of zeste homolog 2 (EZH2) a catalytic component of polycomb repressive complex 2 (PRC2) By inhibiting EZH2 enzymatic activity CPI-169 suppresses methylation at histone H3 lysine 27 (H3K27me3) thereby altering chromatin structure and gene expression profiles Pharmacological experiments demonstrate inhibition of EZH2 with subnanomolar IC50 values and cellular studies indicate reductions of H3K27 trimethylation at nanomolar concentrations CPI-169 has been utilized experimentally to investigate EZH2-dependent epigenetic processes cell proliferation apoptosis pathways and therapeutic potential particularly in lymphoma and cancer biology studies employing in vitro and in vivo models