Learn More
Medchemexpress LLC HY-15846 10mg , CPI-203 CAS:1446144-04-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1584610MG
Encompass_Preferred
HY-15846 10mg CPI-203 CAS: 1446144-04-2 CPI-203 is a novel potent, selective and cell permeable inhibitor of BET bromodomain, with an IC50 value of appr 37 nM (BRD4 α-screen assay). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0955