Learn More
Medchemexpress LLC HY-100421 10mg , CPI-455 CAS:1628208-23-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10042110MG
Encompass_Preferred
HY-100421 10mg CPI-455 CAS: 1628208-23-0 CPI-455 is a specific, pan-KDM5 inhibitor with an IC50 of 10 nM for KDM5A. CPI-455 mediates KDM5 inhibition, elevates global levels of H3K4me3, and decreases the number of drug-tolerant persister cancer cells in multiple cancer cell line models treated with standard chemotherapy or targeted agents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9857