Learn More
Medchemexpress LLC CPI-455 | 1628208-23-0 | 99.0% | 278.31 g/mol | C16H14N4O | 50 MG
SDP
Catalog No. 5000214885
Encompass_Preferred
CPI-455 is a small-molecule pan-KDM5 (histone demethylase) inhibitor with an IC50 of approximately 10 nM for KDM5A. It elevates global H3K4me3 levels and reduces drug-tolerant persister cancer cells in cell-based models. The compound is supplied as a white to off-white solid for research use, with storage and analytical documentation provided by the supplier.
  • Potent KDM5 inhibition (IC50 ≈ 10 nM).
  • Elevates global H3K4 trimethylation (H3K4me3).
  • Demonstrated activity in cell-based models reducing drug-tolerant persister cells.
  • Supplied as a white to off-white solid for in vitro research use only.
  • Analytical purity reported near 99%; confirm lot certificate for exact value.
  • Includes structural identifiers (SMILES) and supporting documentation for characterization.
Catalog No. 50-002-14885 Supplier Medchemexpress LLC Supplier No. HY10042150MG
CPI-455 is a small-molecule pan-KDM5 (histone demethylase) inhibitor with an IC50 of approximately 10 nM for KDM5A. It elevates global H3K4me3 levels and reduces drug-tolerant persister cancer cells in cell-based models. The compound is supplied as a white to off-white solid for research use, with storage and analytical documentation provided by the supplier.
  • Potent KDM5 inhibition (IC50 ≈ 10 nM).
  • Elevates global H3K4 trimethylation (H3K4me3).
  • Demonstrated activity in cell-based models reducing drug-tolerant persister cells.
  • Supplied as a white to off-white solid for in vitro research use only.
  • Analytical purity reported near 99%; confirm lot certificate for exact value.
  • Includes structural identifiers (SMILES) and supporting documentation for characterization.