Learn More
Selleck Chemical LLC CPI-455-5mg
SDP
Catalog No. 502389915
Encompass_Preferred
CPI-455 is a specific KDM5 inhibitor with an IC50 of 10 nM for full-length KDM5A in enzymatic assays. CPI-455 shows over 200-fold selectivity for KDM5 relative to KDM2, 3, 4, 6, and 7 enzymes.
Catalog No. 50-238-9915 Supplier Selleck Chemical LLC Supplier No. S63895MG
CPI-455 is a specific KDM5 inhibitor with an IC50 of 10 nM for full-length KDM5A in enzymatic assays. CPI-455 shows over 200-fold selectivity for KDM5 relative to KDM2, 3, 4, 6, and 7 enzymes.