Learn More
Selleck Chemical LLC CPI-455 HCl 5mg 2095432-28-1
SDP
Catalog No. 501365902
Encompass_Preferred
CPI-455 is a specific KDM5 inhibitor with a half-maximal inhibitory concentration (IC50) of 10 ¿ 1 nM for full-length KDM5A in enzymatic assays, elevating global levels of H3K4 trimethylation (H3K4me3) and decreased the number of DTPs in multiple cancer cell line models treated with standard chemotherapy or targeted agents. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5902 Supplier Selleck Chemical LLC Supplier No. S82875MG
Add to Cart
Add to Cart
CPI-455 is a specific KDM5 inhibitor with a half-maximal inhibitory concentration (IC50) of 10 ¿ 1 nM for full-length KDM5A in enzymatic assays, elevating global levels of H3K4 trimethylation (H3K4me3) and decreased the number of DTPs in multiple cancer cell line models treated with standard chemotherapy or targeted agents. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.