Learn More
Medchemexpress LLC CPI-455 hydrochloride | 2095432-28-1 | 100.0% | 314.77 | C16H15ClN4O | 10 MG
SDP
Catalog No. 5000214002
Encompass_Preferred
CPI-455 hydrochloride is the hydrochloride salt of a potent pan-KDM5 inhibitor used as a research tool to probe KDM5-dependent epigenetic mechanisms. It inhibits KDM5 enzymes (IC50 = 10 nM for KDM5A), elevates H3K4me3 levels in cells, and is supplied with high purity for in vitro studies and mechanism-of-action experiments.
  • Potent KDM5 inhibition (IC50 = 10 nM).
  • Elevates global H3K4me3 levels in cells.
  • High purity for reliable biochemical and cellular assays.
  • Multiple package sizes for experimental flexibility.
  • Recommended storage conditions provided to preserve stability.
  • For research use only; not for clinical or human use.
Catalog No. 50-002-14002 Supplier Medchemexpress LLC Supplier No. HY100421A10MG
CPI-455 hydrochloride is the hydrochloride salt of a potent pan-KDM5 inhibitor used as a research tool to probe KDM5-dependent epigenetic mechanisms. It inhibits KDM5 enzymes (IC50 = 10 nM for KDM5A), elevates H3K4me3 levels in cells, and is supplied with high purity for in vitro studies and mechanism-of-action experiments.
  • Potent KDM5 inhibition (IC50 = 10 nM).
  • Elevates global H3K4me3 levels in cells.
  • High purity for reliable biochemical and cellular assays.
  • Multiple package sizes for experimental flexibility.
  • Recommended storage conditions provided to preserve stability.
  • For research use only; not for clinical or human use.