Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CPI-455 hydrochloride | 2095432-28-1 | 100.0% | 314.77 g/mol | C16H15ClN4O | 5 MG
SDP

Catalog No. 5000213937
Encompass_Preferred

CPI-455 hydrochloride is the hydrochloride salt of a potent, selective pan-KDM5 histone demethylase inhibitor used in biochemical and cellular epigenetic research. It inhibits KDM5A with an IC50 of ≈10 nM, elevates global H3K4 trimethylation, and has been applied in studies of drug-tolerant persister cancer cells.

  • Potent pan-KDM5 inhibitor (IC50 ≈ 10 nM for KDM5A).
  • Elevates global H3K4 trimethylation in cellular assays.
  • Hydrochloride salt form for enhanced stability and handling.
  • Molecular formula C16H15ClN4O; molecular weight 314.77 g/mol.
  • CAS number 2095432-28-1 for unambiguous identification.
  • High purity (listed as 99.95%), suitable for research applications.
  • Soluble in DMSO; water insoluble.
  • Available in small laboratory pack sizes for screening and mechanistic studies.

Catalog No. 50-002-13937 Supplier Medchemexpress LLC Supplier No. HY100421A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CPI-455 hydrochloride is the hydrochloride salt of a potent, selective pan-KDM5 histone demethylase inhibitor used in biochemical and cellular epigenetic research. It inhibits KDM5A with an IC50 of ≈10 nM, elevates global H3K4 trimethylation, and has been applied in studies of drug-tolerant persister cancer cells.

  • Potent pan-KDM5 inhibitor (IC50 ≈ 10 nM for KDM5A).
  • Elevates global H3K4 trimethylation in cellular assays.
  • Hydrochloride salt form for enhanced stability and handling.
  • Molecular formula C16H15ClN4O; molecular weight 314.77 g/mol.
  • CAS number 2095432-28-1 for unambiguous identification.
  • High purity (listed as 99.95%), suitable for research applications.
  • Soluble in DMSO; water insoluble.
  • Available in small laboratory pack sizes for screening and mechanistic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.