Learn More
Selleck Chemical LLC Devimistat (CPI-613) 5mg 95809-78-2
SDP
Catalog No. 501362936
Encompass_Preferred
Devimistat (CPI-613), a lipoate analog, inhibits mitochondrial enzymes pyruvate dehydrogenase (PDH) and ?-ketoglutarate dehydrogenase in NCI-H460 cell line, disrupts tumor cell mitochondrial metabolism. CPI-613 induces apoptosis in pancreatic cancer cells. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2936 Supplier Selleck Chemical LLC Supplier No. S27765MG
Devimistat (CPI-613), a lipoate analog, inhibits mitochondrial enzymes pyruvate dehydrogenase (PDH) and ?-ketoglutarate dehydrogenase in NCI-H460 cell line, disrupts tumor cell mitochondrial metabolism. CPI-613 induces apoptosis in pancreatic cancer cells. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.