Learn More
TARGETMOL CHEMICALS INC CPI-637 10MG
SDP
Catalog No. 502876909
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. CPI-637 is a selective and cell-active benzodiazepinone CBP/EP300 bromodomain inhibitor. Purity 99.91%
Catalog No. 50-287-6909 Supplier TARGETMOL CHEMICALS INC Supplier No. T681110MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. CPI-637 is a selective and cell-active benzodiazepinone CBP/EP300 bromodomain inhibitor. Purity 99.91%