Learn More
Selleck Chemical LLC CPI-637
SDP
Catalog No. 501365775
Encompass_Preferred
CPI-637 is a selective and cell-active benzodiazepinone CBP/EP300 bromodomain inhibitor with IC50 values of 0 03 M and 0 051 M for CBP and EP300 respectively in TR-FRET assay It is highly selective against other bromodomains displaying substantial biochemical activity only against BRD9
Catalog No. 50-136-5775 Supplier Selleck Chemical LLC Supplier No. S819010MG
CPI-637 is a selective and cell-active benzodiazepinone CBP/EP300 bromodomain inhibitor with IC50 values of 0 03 M and 0 051 M for CBP and EP300 respectively in TR-FRET assay It is highly selective against other bromodomains displaying substantial biochemical activity only against BRD9