Learn More
Medchemexpress LLC CPI-637 | 1884712-47-3 | MFCD30489741 | 99.9% | 386.45 g/mol | C22H22N6O | 10 MG
SDP
Catalog No. 5000214983
Encompass_Preferred
CPI-637 is a selective, cell-active CBP/EP300 bromodomain inhibitor used in epigenetic and cancer biology research. It demonstrates low-nanomolar biochemical potency against CBP and EP300 and shows cellular activity in the submicromolar range.
  • Selective inhibition of CBP and EP300 bromodomains (IC50 = 0.03 μM and 0.051 μM).
  • Reduced activity against BRD4 BD1 (IC50 = 11.0 μM).
  • Cellular activity with CBP BRET EC50 ≈ 0.3 μM.
  • High reported purity (~99.9%) suitable for research applications.
  • Chemical formula C22H22N6O; molecular weight 386.45 g·mol⁻¹.
Catalog No. 50-002-14983 Supplier Medchemexpress LLC Supplier No. HY10048210MG
CPI-637 is a selective, cell-active CBP/EP300 bromodomain inhibitor used in epigenetic and cancer biology research. It demonstrates low-nanomolar biochemical potency against CBP and EP300 and shows cellular activity in the submicromolar range.
  • Selective inhibition of CBP and EP300 bromodomains (IC50 = 0.03 μM and 0.051 μM).
  • Reduced activity against BRD4 BD1 (IC50 = 11.0 μM).
  • Cellular activity with CBP BRET EC50 ≈ 0.3 μM.
  • High reported purity (~99.9%) suitable for research applications.
  • Chemical formula C22H22N6O; molecular weight 386.45 g·mol⁻¹.