Learn More
TARGETMOL CHEMICALS INC CPI203 25MG
SDP
Catalog No. 502577120
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CPI203 (CPI 203) is an effective BET bromodomain inhibitor (IC50 37 nM for BRD4). purity: 99%
Catalog No. 50-257-7120 Supplier TARGETMOL CHEMICALS INC Supplier No. T244225MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CPI203 (CPI 203) is an effective BET bromodomain inhibitor (IC50 37 nM for BRD4). purity: 99%