Learn More
Medchemexpress LLC Cpk20 | 2397708-22-2 | 99.7% | 506.50 | 25 MG
SDP
Catalog No. 5000394300
Encompass_Preferred
CPK20 is a potent and highly selective KCNT1 channel blocker. It is intended for research use only and not for sale to patients.
  • Potent and highly selective KCNT1 channel blocker.
  • Can be used in the study of epilepsy.
Catalog No. 50-003-94300 Supplier Medchemexpress LLC Supplier No. 25MGHY162549
CPK20 is a potent and highly selective KCNT1 channel blocker. It is intended for research use only and not for sale to patients.
  • Potent and highly selective KCNT1 channel blocker.
  • Can be used in the study of epilepsy.