Learn More
Cell Systems Corporation Complete Medium Kit With Serum and CultureBoost-R™ (4Z0-500-R)
SDP
Catalog No. NC0039601
Encompass_Preferred
The Complete Medium Kit With Serum and CultureBoost-R is formulated with 10% serum and becomes a complete medium once activated with the included CultureBoost-R supplement. This kit consists of 500mL Cell Systems Medium (4Z0-500-R), 10mL CultureBoost-R (containing human recombinant growth factors) and 10mL Attachment Factor (extracellular matrix reagent for coating flasks). ). 4Z0-500-R is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.
Catalog No. NC0039601 Supplier Cell Systems Corporation Supplier No. 4Z0500R
The Complete Medium Kit With Serum and CultureBoost-R is formulated with 10% serum and becomes a complete medium once activated with the included CultureBoost-R supplement. This kit consists of 500mL Cell Systems Medium (4Z0-500-R), 10mL CultureBoost-R (containing human recombinant growth factors) and 10mL Attachment Factor (extracellular matrix reagent for coating flasks). ). 4Z0-500-R is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.