Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier:  Thermo Scientific 6041.5730
Encompass
- Virtually zero-dead-volume connections ensure optimal separation efficiencies - Preassembled and finger-tightened for easy tool-free instrument setup and configuration - High-pressure tested to provide UHPLC compatibility up to 1200 bar (17400 psi) - Suitable for temperatures up to 120C - Flexible PEEK sheathed fused silica capillaries 1/32 in OD - Compatible with valves and columns from virtually any manufacturer
Catalog No. 50-134-7932