Learn More
Trajan Scientific CPLRY 15QC2.5/BPX50-0.25
SDP
Catalog No. 502530233
Encompass_Preferred
SGE BPX50 Capillary Column, Film Thickness: 0.25 um, Particle Size: 0.25 um, Length: 49 ft., 15 m, Diameter Inner: 0.25 mm, For Use With: Drug Screening, GC, Steroids Analysis, Herbicides Analysis, Packing Material: SGE BPX50, Stationary Phase: 50% Phenyl Polysilphenylene Siloxane
Catalog No. 50-253-0233 Supplier Trajan Scientific Supplier No. 054750
SGE BPX50 Capillary Column, Film Thickness: 0.25 um, Particle Size: 0.25 um, Length: 49 ft., 15 m, Diameter Inner: 0.25 mm, For Use With: Drug Screening, GC, Steroids Analysis, Herbicides Analysis, Packing Material: SGE BPX50, Stationary Phase: 50% Phenyl Polysilphenylene Siloxane