Learn More
Trajan Scientific CPLRY 30M BP5 0.25UM EA
SDP
Catalog No. 502530239
Encompass_Preferred
SGE BP5 Capillary GC Column, Diameter Inner: 0.25 mm, For Use With: Hydrocarbon Analysis, Herbicides Analysis, GC, Drugs of Abuse, Pesticide Analysis, Film Thickness: 0.25 um, Length: 30 m, 98 ft., Packing Material: SGE BP5, Particle Size: 0.25 um
Catalog No. 50-253-0239 Supplier Trajan Scientific Supplier No. 054183
SGE BP5 Capillary GC Column, Diameter Inner: 0.25 mm, For Use With: Hydrocarbon Analysis, Herbicides Analysis, GC, Drugs of Abuse, Pesticide Analysis, Film Thickness: 0.25 um, Length: 30 m, 98 ft., Packing Material: SGE BP5, Particle Size: 0.25 um