Learn More
Trajan Scientific CPLRY 30M X 0.53MMID SOLGEL-WA
SDP
Catalog No. 502530232
Encompass_Preferred
Trajan SolGel WAX Columns, Diameter Inner: 0.53 mm, Film Thickness: 1 um, Length: 30 m, For Use With: Phenols, Sugars-alditol Acetates, Food - FAME, Essential Oils, Acids, Alcoholic Beverages Analysis, Amines Aliphatic, Amines Aromatic, Packing Material: SolGel-Wax
Catalog No. 50-253-0232 Supplier Trajan Scientific Supplier No. 054785
Trajan SolGel WAX Columns, Diameter Inner: 0.53 mm, Film Thickness: 1 um, Length: 30 m, For Use With: Phenols, Sugars-alditol Acetates, Food - FAME, Essential Oils, Acids, Alcoholic Beverages Analysis, Amines Aliphatic, Amines Aromatic, Packing Material: SolGel-Wax