Learn More
Trajan Scientific CPLRY 50QC3/BP1-1.0
SDP
Catalog No. 502530244
Encompass_Preferred
Trajan BP1/BP1 PONA Column, Diameter Inner: 0.32 mm, Film Thickness: 1 um, Length: 50 m, For Use With: Aromatics Analysis, Herbicides Analysis, Phenols, Hydrocarbon Analysis, Pesticide Analysis, Amines, Stationary Phase: 100% Dimethyl Polysiloxane, Polarity: Non polar
Catalog No. 50-253-0244 Supplier Trajan Scientific Supplier No. 054072
Trajan BP1/BP1 PONA Column, Diameter Inner: 0.32 mm, Film Thickness: 1 um, Length: 50 m, For Use With: Aromatics Analysis, Herbicides Analysis, Phenols, Hydrocarbon Analysis, Pesticide Analysis, Amines, Stationary Phase: 100% Dimethyl Polysiloxane, Polarity: Non polar