Learn More
Trajan Scientific CPLRY BP20-30MX0.25MMX1UM
SDP
Catalog No. 502530237
Encompass_Preferred
SGE BP20 (WAX) Capillary Columns, Diameter Inner: 0.25 mm, Film Thickness: 1 um, Length: 30 m, For Use With: GC, Fragrance Analysis, Aldehydes Analysis, Ketones, Packing Material: SGE BP20 (Wax), Particle Size: 1 um, Stationary Phase: Polyethylene Glycol, Polarity: Polar
Catalog No. 50-253-0237 Supplier Trajan Scientific Supplier No. 054439
SGE BP20 (WAX) Capillary Columns, Diameter Inner: 0.25 mm, Film Thickness: 1 um, Length: 30 m, For Use With: GC, Fragrance Analysis, Aldehydes Analysis, Ketones, Packing Material: SGE BP20 (Wax), Particle Size: 1 um, Stationary Phase: Polyethylene Glycol, Polarity: Polar