Learn More
Trajan Scientific CPLRY BPX1-6MX0.53MMX2.65UM
SDP
Catalog No. 502530229
Encompass_Preferred
SGE GC Capillary Columns: Dimethyl Polysiloxane, Diameter Inner: 0.53 mm, Film Thickness: 2.65 um, Length: 6 m, Temperature Range: -30 deg.C to +370 deg.C, For Use With: Simulated Distillation, Packing Material: BPX1, Particle Size: 2.65 um, Stationary Phase: 100% Dimethyl Polysiloxane
Catalog No. 50-253-0229 Supplier Trajan Scientific Supplier No. 0548025
SGE GC Capillary Columns: Dimethyl Polysiloxane, Diameter Inner: 0.53 mm, Film Thickness: 2.65 um, Length: 6 m, Temperature Range: -30 deg.C to +370 deg.C, For Use With: Simulated Distillation, Packing Material: BPX1, Particle Size: 2.65 um, Stationary Phase: 100% Dimethyl Polysiloxane