Learn More
Trajan Scientific CPLRY BPX5-15MX0.25MMX0.25UM
SDP
Catalog No. 502530243
Encompass_Preferred
Trajan BPX5 GC Capillary Columns, Diameter Inner: 0.25 mm, Film Thickness: 0.25 um, Length: 15 m, For Use With: Hydrocarbons, Pesticides and Herbicides Analysis, GC/MS, Amines, Solvents Analysis, Phenols, Ultra Trace Analyses, Packing Material: BPX5
Catalog No. 50-253-0243 Supplier Trajan Scientific Supplier No. 054100
Add to Cart
Add to Cart
Trajan BPX5 GC Capillary Columns, Diameter Inner: 0.25 mm, Film Thickness: 0.25 um, Length: 15 m, For Use With: Hydrocarbons, Pesticides and Herbicides Analysis, GC/MS, Amines, Solvents Analysis, Phenols, Ultra Trace Analyses, Packing Material: BPX5