Learn More
Trajan Scientific CPLRY BPX50-10MX0.1MMX0.1UM
SDP
Catalog No. 502530234
Encompass_Preferred
SGE BPX50 Capillary Column, Film Thickness: 0.1 um, Particle Size: 0.1 um, Length: 33 ft., 10 m, Diameter Inner: 0.1 mm, For Use With: Triazines, Steroids Analysis, Drug Screening, Herbicides Analysis, Packing Material: SGE BPX50, Stationary Phase: 50% Phenyl Polysilphenylene Siloxane
Catalog No. 50-253-0234 Supplier Trajan Scientific Supplier No. 054740
SGE BPX50 Capillary Column, Film Thickness: 0.1 um, Particle Size: 0.1 um, Length: 33 ft., 10 m, Diameter Inner: 0.1 mm, For Use With: Triazines, Steroids Analysis, Drug Screening, Herbicides Analysis, Packing Material: SGE BPX50, Stationary Phase: 50% Phenyl Polysilphenylene Siloxane