Learn More
Trajan Scientific CPLRY COL 50QC1.5/BP1 PONA
SDP
Catalog No. 502530227
Encompass_Preferred
Trajan BP1 Pona GC Capillary Columns, Diameter Inner: 0.15 mm, Film Thickness: 0.5 um, Length: 50 m, Max. Temperature: 320/340 deg.C
Catalog No. 50-253-0227 Supplier Trajan Scientific Supplier No. 054950
Trajan BP1 Pona GC Capillary Columns, Diameter Inner: 0.15 mm, Film Thickness: 0.5 um, Length: 50 m, Max. Temperature: 320/340 deg.C