Learn More
Trajan Scientific CPLRY COL 50QC2/CYDEX B-0.25
SDP
Catalog No. 502530228
Encompass_Preferred
Trajan Cydex-B GC Capillary Columns, Diameter Inner: 0.22 mm, Film Thickness: 0.25 um, Length: 25 m, Max. Temperature: 220/240 deg.C, Cydex-B GC Column
Catalog No. 50-253-0228 Supplier Trajan Scientific Supplier No. 054901
Trajan Cydex-B GC Capillary Columns, Diameter Inner: 0.22 mm, Film Thickness: 0.25 um, Length: 25 m, Max. Temperature: 220/240 deg.C, Cydex-B GC Column