Learn More
Aviva Systems Biology Corp CPLX2 Antibody - N-terminal region (ARP52225_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52225_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CPLX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CPLX2. Alternate names: 921-L, CPX-2, CPX2, Hfb1, MGC138492ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-664