Learn More
Sino Biological CPLX2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11927T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CPLX2 (rh CPLX2; Catalog#11927-H07E; Q6PUV4-1; Asp2-Lys134). CPLX2 specific IgG was purified by Human CPLX2 affinity chromatography.
Catalog No. 50-224-6382