Learn More
Biotium CPM
Catalog No. 501964828
Encompass_Preferred
CPM (full name: 7-diethylamino-3-(4'-maleimidylphenyl)-4-methylcoumarin) is probably the most widely used blue fluorescent thiol-reactive dye. This maleimide derivative of coumarin is essentially nonfluorescent until it reacts with thiols, making it possible to quantify thiols without a separation step. CPM is a good energy acceptor from tryptophan and a good energy donor to fluorescein.
Catalog No. 50-196-4828 Supplier Biotium Supplier No. 91010
CPM (full name: 7-diethylamino-3-(4'-maleimidylphenyl)-4-methylcoumarin) is probably the most widely used blue fluorescent thiol-reactive dye. This maleimide derivative of coumarin is essentially nonfluorescent until it reacts with thiols, making it possible to quantify thiols without a separation step. CPM is a good energy acceptor from tryptophan and a good energy donor to fluorescein.