Learn More
Medchemexpress LLC Cpn-219 | 2240202-02-0 | 99.1% | 849.08 | 1 MG
SDP
Catalog No. 5000398637
Encompass_Preferred
CPN-219 is a next-generation hexapeptidic NMUR2 agonist that achieves a dose-dependent and selective activation of NMUR2 with an EC50 value of 2.2 nM. It is for research use only.
  • Acts as a next-generation hexapeptidic NMUR2 agonist
  • Achieves dose-dependent and selective activation of NMUR2
  • Demonstrates an EC50 value of 2.2 nM in CHO cells for NMUR2 activation
  • Suppresses body weight gain in mice via intranasal administration
  • Soluble in H₂O and DMSO at 50 mg/mL
Catalog No. 50-003-98637 Supplier Medchemexpress LLC Supplier No. HYP30301MG
Add to Cart
Add to Cart
CPN-219 is a next-generation hexapeptidic NMUR2 agonist that achieves a dose-dependent and selective activation of NMUR2 with an EC50 value of 2.2 nM. It is for research use only.
  • Acts as a next-generation hexapeptidic NMUR2 agonist
  • Achieves dose-dependent and selective activation of NMUR2
  • Demonstrates an EC50 value of 2.2 nM in CHO cells for NMUR2 activation
  • Suppresses body weight gain in mice via intranasal administration
  • Soluble in H₂O and DMSO at 50 mg/mL