Learn More
Medchemexpress LLC CPN-351 TFA | 912.11 (free base) | 5 MG
SDP
Catalog No. 5000423784
Encompass_Preferred
CPN-351 TFA, a pentapeptide, acts as a selective antagonist for human NMUR1, demonstrating a pA2 of 7.35. Its antagonistic effect on human NMUR1 is significantly higher, specifically 10 times greater, than its effect on NMUR2. This compound is suitable for application in inflammation research.
  • Selective antagonist of human NMUR1.
  • Pentapeptide compound.
  • Antagonistic effect on human NMUR1 is 10 times higher than on NMUR2.
  • Used in the study of inflammation.
Catalog No. 50-004-23784 Supplier Medchemexpress LLC Supplier No. HYP10473A5MG
CPN-351 TFA, a pentapeptide, acts as a selective antagonist for human NMUR1, demonstrating a pA2 of 7.35. Its antagonistic effect on human NMUR1 is significantly higher, specifically 10 times greater, than its effect on NMUR2. This compound is suitable for application in inflammation research.
  • Selective antagonist of human NMUR1.
  • Pentapeptide compound.
  • Antagonistic effect on human NMUR1 is 10 times higher than on NMUR2.
  • Used in the study of inflammation.